COMH42117438 Combi-Blocks, Inc.

T-Boc-n-amido-peg12-propargyl

  • MDL number MFCD31615451
  • Cas Number : 1520979-34-3
  • Molecular Weight : 683.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4058
Availability details for
Quantity
  • Availability
  • Add to Cart