COMH42117443 Combi-Blocks, Inc.

1-Benzyl-2-oxo-1,2-dihydro-quinoline-4-carbaldehyde

  • MDL number MFCD22385564
  • Cas Number : 854857-64-0
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4125
Availability details for
Quantity
  • Availability
  • Add to Cart