COMH42117445 Combi-Blocks, Inc.

[3-(4-Chlorophenyl)-5,9-dimethyl-7-oxo-7h-furo[3,2-g]chromen-6-yl]acetic acid

  • MDL number MFCD03661308
  • Cas Number : 664366-16-9
  • Molecular Weight : 382.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4132
Availability details for
Quantity
  • Availability
  • Add to Cart