COMH4211744C Combi-Blocks, Inc.

1-(6-Chloropyridazin-3-yl)piperidin-4-amine dihydrochloride

  • MDL number MFCD27756343
  • Cas Number : 1984117-56-7
  • Molecular Weight : 285.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4154
Availability details for
Quantity
  • Availability
  • Add to Cart