COMH4211744E Combi-Blocks, Inc.

Dbco-nhco-peg13-nhs ester

  • MDL number MFCD29052180
  • Molecular Weight : 1046.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4165
Availability details for
Quantity
  • Availability
  • Add to Cart