COMH42117450 Combi-Blocks, Inc.

2-Bromo-3-fluoro-6-methoxybenzaldehyde

  • MDL number MFCD31619542
  • Cas Number : 1780200-30-7
  • Molecular Weight : 233.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4192
Availability details for
Quantity
  • Availability
  • Add to Cart