COMH42117452 Combi-Blocks, Inc.

2-[2-(1,3-Dioxo-1,3-dihydro-2h-isoindol-2-yl)ethoxy]isonicotinonitrile

  • MDL number MFCD31583438
  • Cas Number : 2270909-82-3
  • Molecular Weight : 293.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-(1,3-dioxoisoindolin-2-yl)ethoxy)isonicotinonitrile
Inchi Code 1S/C16H11N3O3/c17-10-11-5-6-18-14(9-11)22-8-7-19-15(20)12-3-1-2-4-13(12)16(19)21/h1-6,9H,7-8H2
Inchi Key QTKGOLHHAZDJIZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4194
Availability details for
Quantity
  • Availability
  • Add to Cart