COMH42117456 Combi-Blocks, Inc.

1-Dimethylamino-2,3-epoxypropane

  • MDL number MFCD09740456
  • Cas Number : 13222-40-7
  • Molecular Weight : 101.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4201
Availability details for
Quantity
  • Availability
  • Add to Cart