COMH4211745F Combi-Blocks, Inc.

4-[(4-Butoxyphenyl)diazenyl]benzenesulfonic acid

  • MDL number MFCD31653106
  • Molecular Weight : 334.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4236
Availability details for
Quantity
  • Availability
  • Add to Cart