COMH42117463 Combi-Blocks, Inc.

N-(T-Butyl ester-peg3)-n-bis(peg3-amine)

  • MDL number MFCD31561124
  • Cas Number : 2183440-29-9
  • Molecular Weight : 627.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4253
Availability details for
Quantity
  • Availability
  • Add to Cart