COMH42117466 Combi-Blocks, Inc.

N2,N2-Dimethyl-n1-(2-piperidin-4-ylethyl)glycinamide dihydrochloride

  • MDL number MFCD31537587
  • Cas Number : 2197685-83-7
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4265
Availability details for
Quantity
  • Availability
  • Add to Cart