COMH42117470 Combi-Blocks, Inc.

1-(2-Phenoxypyridin-3-yl)methanamine dihydrochloride

  • MDL number MFCD15173360
  • Cas Number : 1158627-80-5
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4317
Availability details for
Quantity
  • Availability
  • Add to Cart