COMH42117474 Combi-Blocks, Inc.

2-[4-(1-Adamantyl)phenoxy]ethanamine hydrochloride

  • MDL number MFCD07282581
  • Cas Number : 1052545-53-5
  • Molecular Weight : 307.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4331
Availability details for
Quantity
  • Availability
  • Add to Cart