COMH4211747B Combi-Blocks, Inc.

M-Peg6-(ch2)8-phosphonic acid

  • MDL number MFCD30458009
  • Cas Number : 2028284-71-9
  • Molecular Weight : 444.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4373
Availability details for
Quantity
  • Availability
  • Add to Cart