COMH4211747D Combi-Blocks, Inc.

2-Methyl-4-nitro-2h-1,2,3-triazole

  • MDL number MFCD31583429
  • Cas Number : 107945-73-3
  • Molecular Weight : 128.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4381

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4381
Availability details for
Quantity
  • Availability
  • Add to Cart