COMH42117483 Combi-Blocks, Inc.

4-(2,4-Dichlorophenoxy)aniline hydrochloride

  • MDL number MFCD27961518
  • Cas Number : 72782-76-4
  • Molecular Weight : 290.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4406

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4406
Availability details for
Quantity
  • Availability
  • Add to Cart