COMH4211748B Combi-Blocks, Inc.

2-Amino-n-cyclopropyl-3-methylbenzamide

  • MDL number MFCD09050748
  • Cas Number : 926197-46-8
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4454
Availability details for
Quantity
  • Availability
  • Add to Cart