COMH4211748C Combi-Blocks, Inc.

N,N'-Dimethylbenzene-1,2-diamine dihydrochloride

  • MDL number MFCD31583442
  • Cas Number : 75960-46-2
  • Molecular Weight : 209.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3843
Availability details for
Quantity
  • Availability
  • Add to Cart