COMH421174A7 Combi-Blocks, Inc.

(1R)-(-)-(10-Camphorsulfonyl)oxaziridine

  • MDL number MFCD30486441
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4aS,7S,8aR)-9,9-dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide
Inchi Code 1S/C10H15NO3S/c1-8(2)7-3-4-9(8)6-15(12,13)11-10(9,5-7)14-11/h7H,3-6H2,1-2H3/t7-,9-,10+,11?/m0/s1
Inchi Key GBBJBUGPGFNISJ-ASMZKLSYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4598
Availability details for
Quantity
  • Availability
  • Add to Cart