COMH421174A8 Combi-Blocks, Inc.

1-Benzyl-4-[(3,5-dimethyl-1h-pyrazol-1-yl)methyl]piperazine

  • MDL number MFCD01764172
  • Cas Number : 278782-31-3
  • Molecular Weight : 284.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4599
Availability details for
Quantity
  • Availability
  • Add to Cart