COMH421174A9 Combi-Blocks, Inc.

8-Chloro-2,3,4,9-tetrahydro-1h-carbazol-1-amine

  • MDL number MFCD11168346
  • Cas Number : 1042626-75-4
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4603
Availability details for
Quantity
  • Availability
  • Add to Cart