COMH421174AF Combi-Blocks, Inc.

3-Heptadecyl-1h-1,2,4-triazol-5-amine

  • MDL number MFCD31619046
  • Cas Number : 854738-04-8
  • Molecular Weight : 322.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-heptadecyl-1H-1,2,4-triazol-5-amine
Inchi Code 1S/C19H38N4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-21-19(20)23-22-18/h2-17H2,1H3,(H3,20,21,22,23)
Inchi Key QQGWGESQLZHLOI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4625

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4625
Availability details for
Quantity
  • Availability
  • Add to Cart