COMH421174B2 Combi-Blocks, Inc.

Dspe-peg8-mal

  • MDL number MFCD30828705
  • Cas Number : 2112737-94-5
  • Molecular Weight : 1251.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4661
Availability details for
Quantity
  • Availability
  • Add to Cart