COMH421174B9 Combi-Blocks, Inc.

1-Acetyl-2-thiohydantoin

  • MDL number MFCD00020879
  • Cas Number : 584-26-9
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4698
Availability details for
Quantity
  • Availability
  • Add to Cart