COMH421174BA Combi-Blocks, Inc.

6,8-Difluoro-2,2,4-trimethyl-1,2-dihydro-quinoline

  • MDL number MFCD15525558
  • Cas Number : 1260877-83-5
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6,8-difluoro-2,2,4-trimethyl-1,2-dihydroquinoline
Inchi Code 1S/C12H13F2N/c1-7-6-12(2,3)15-11-9(7)4-8(13)5-10(11)14/h4-6,15H,1-3H3
Inchi Key YRLLYIALIGRLDY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4700
Availability details for
Quantity
  • Availability
  • Add to Cart