COMH421174C1 Combi-Blocks, Inc.

3-Cyclopropyl-1-methyl-1h-pyrazole-5-carboxamide

  • MDL number MFCD09701848
  • Cas Number : 1172483-17-8
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4733
Availability details for
Quantity
  • Availability
  • Add to Cart