COMH421174C2 Combi-Blocks, Inc.

Ethyl (3-hydroxy-1h-pyrazol-4-yl)acetate

  • MDL number MFCD09034162
  • Cas Number : 890095-13-3
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4743
Availability details for
Quantity
  • Availability
  • Add to Cart