COMH421174C3 Combi-Blocks, Inc.

(S)-2-(3-Cyclopropylureido)-2-phenylacetic acid

  • MDL number MFCD22573473
  • Cas Number : 1401320-37-3
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-2-(3-cyclopropylureido)-2-phenylacetic acid
Inchi Code 1S/C12H14N2O3/c15-11(16)10(8-4-2-1-3-5-8)14-12(17)13-9-6-7-9/h1-5,9-10H,6-7H2,(H,15,16)(H2,13,14,17)/t10-/m0/s1
Inchi Key WRVJRYBZXIWNKR-JTQLQIEISA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4748
Availability details for
Quantity
  • Availability
  • Add to Cart