COMH421174C9 Combi-Blocks, Inc.

2,5-Dioxopyrrolidin-1-yl 3-(2-((tert-butoxycarbonyl)amino)ethoxy)propanoate

  • MDL number MFCD20227793
  • Cas Number : 1260092-55-4
  • Molecular Weight : 330.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4779
Availability details for
Quantity
  • Availability
  • Add to Cart