COMH421174D4 Combi-Blocks, Inc.

7-(Diethylamino)-3-nitro-2h-chromen-2-one

  • MDL number MFCD27966252
  • Cas Number : 118116-71-5
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4801
Availability details for
Quantity
  • Availability
  • Add to Cart