COMH421174D7 Combi-Blocks, Inc.

1-([1,2,4]Triazolo[4,3-a]pyridin-3-yl)-3-methylbutan-1-amine hydrochloride

  • MDL number MFCD18483731
  • Cas Number : 1179491-42-9
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4834
Availability details for
Quantity
  • Availability
  • Add to Cart