COMH421174DD Combi-Blocks, Inc.

1-(Methylamino)-3-piperidin-1-ylpropan-2-ol

  • MDL number MFCD11108828
  • Cas Number : 1019527-68-4
  • Molecular Weight : 172.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4846

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4846
Availability details for
Quantity
  • Availability
  • Add to Cart