COMH421174E1 Combi-Blocks, Inc.

1-[3-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD03988235
  • Cas Number : 63674-52-2
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4863
Availability details for
Quantity
  • Availability
  • Add to Cart