COMH421174E3 Combi-Blocks, Inc.

(4-Bromophenyl)(5-hydroxy-1-benzofuran-3-yl)methanone

  • MDL number MFCD05739936
  • Cas Number : 723247-35-6
  • Molecular Weight : 317.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4885

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4885
Availability details for
Quantity
  • Availability
  • Add to Cart