COMH421174E8 Combi-Blocks, Inc.

[(Diphenylmethyl)sulfanyl]methanimidamide hydrochloride

  • MDL number MFCD31583444
  • Cas Number : 60282-85-1
  • Molecular Weight : 278.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzhydryl carbamimidothioate hydrochloride
Inchi Code 1S/C14H14N2S.ClH/c15-14(16)17-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10,13H,(H3,15,16);1H
Inchi Key IQFNCNHJOCSBHF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4892
Availability details for
Quantity
  • Availability
  • Add to Cart