COMH421174EE Combi-Blocks, Inc.

1'-[(tert-Butoxycarbonyl)amino]-1,1'-bi(cyclopropyl)-1-carboxylic acid

  • MDL number MFCD28642664
  • Cas Number : 1784118-69-9
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1'-((tert-butoxycarbonyl)amino)-[1,1'-bi(cyclopropane)]-1-carboxylic acid
Inchi Code 1S/C12H19NO4/c1-10(2,3)17-9(16)13-12(6-7-12)11(4-5-11)8(14)15/h4-7H2,1-3H3,(H,13,16)(H,14,15)
Inchi Key KLLSEJPLBJVYTB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4919
Availability details for
Quantity
  • Availability
  • Add to Cart