COMH421174F1 Combi-Blocks, Inc.

Boc-l-ser(bzl)-o-ch2-f-ch2-cooh dcha

  • MDL number MFCD30749175
  • Cas Number : 1221293-89-5
  • Molecular Weight : 624.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4927
Availability details for
Quantity
  • Availability
  • Add to Cart