COMH421174F2 Combi-Blocks, Inc.

3-[(4-Fluorophenyl)methylidene]pentane-2,4-dione

  • MDL number MFCD00488973
  • Cas Number : 15851-94-2
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4944
Availability details for
Quantity
  • Availability
  • Add to Cart