COMH42117502 Combi-Blocks, Inc.

Fmoc-nh-ethyl-ss-propionic nhs ester

  • MDL number MFCD31536765
  • Cas Number : 2128735-23-7
  • Molecular Weight : 500.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5030
Availability details for
Quantity
  • Availability
  • Add to Cart