COMH42117503 Combi-Blocks, Inc.

3-Anilino-2-(3,4,5-trimethoxybenzyl)acrylonitrile

  • MDL number MFCD09998794
  • Cas Number : 118791-61-0
  • Molecular Weight : 324.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5032
Availability details for
Quantity
  • Availability
  • Add to Cart