COMH42117506 Combi-Blocks, Inc.

6-Amino-2-[(2,4-difluorophenyl)amino]pyrimidin-4(3h)-one

  • MDL number MFCD31581339
  • Cas Number : 123375-87-1
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5035
Availability details for
Quantity
  • Availability
  • Add to Cart