COMH42117508 Combi-Blocks, Inc.

2-[(1E)-(2-Chloropyridin-3-yl)methylene]aminoethanol

  • MDL number MFCD31583433
  • Cas Number : 2270915-07-4
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5054

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5054
Availability details for
Quantity
  • Availability
  • Add to Cart