COMH42117509 Combi-Blocks, Inc.

Methyl (1r,2r,3r,4r)-3-(aminocarbonyl)bicyclo[2.2.1]hept-5-ene-2-carboxylate

  • MDL number MFCD31583415
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5065
Availability details for
Quantity
  • Availability
  • Add to Cart