COMH42117516 Combi-Blocks, Inc.

2-Fluoro-3-methyl-5-nitroanisole

  • MDL number MFCD28797444
  • Cas Number : 1804495-50-8
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5141
Availability details for
Quantity
  • Availability
  • Add to Cart