COMH4211751D Combi-Blocks, Inc.

M-Peg6-(ch2)8-phosphonic acid ethyl ester

  • MDL number MFCD30458010
  • Cas Number : 2028281-88-9
  • Molecular Weight : 500.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5164
Availability details for
Quantity
  • Availability
  • Add to Cart