COMH42117521 Combi-Blocks, Inc.

(4-Oxo-2-phenyl-1,4-dihydroquinazolin-3(2h)-yl)acetic acid

  • MDL number MFCD18447000
  • Cas Number : 1269529-74-9
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5187
Availability details for
Quantity
  • Availability
  • Add to Cart