COMH42117522 Combi-Blocks, Inc.

Amino-peg11-alcohol

  • MDL number MFCD31561149
  • Cas Number : 15332-94-2
  • Molecular Weight : 501.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5190
Availability details for
Quantity
  • Availability
  • Add to Cart