COMH42117528 Combi-Blocks, Inc.

3-Bromo-1-(3,4-dichlorophenyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD31581334
  • Cas Number : 2186672-66-0
  • Molecular Weight : 321.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5218

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5218
Availability details for
Quantity
  • Availability
  • Add to Cart