COMH4211752B Combi-Blocks, Inc.

Hydroxy-peg4-nitrile

  • MDL number MFCD31615437
  • Cas Number : 2194563-83-0
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5228
Availability details for
Quantity
  • Availability
  • Add to Cart