COMH42117530 Combi-Blocks, Inc.

2-(Aminomethyl)-1-(cyclopropylcarbonyl)-3,5-dimethylpyridin-4(1h)-one hydrochloride

  • MDL number MFCD31557143
  • Cas Number : 2270906-61-9
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(aminomethyl)-1-(cyclopropanecarbonyl)-3,5-dimethylpyridin-4(1H)-one hydrochloride
Inchi Code 1S/C12H16N2O2.ClH/c1-7-6-14(12(16)9-3-4-9)10(5-13)8(2)11(7)15;/h6,9H,3-5,13H2,1-2H3;1H
Inchi Key SNTODUHHTULCIL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5260
Availability details for
Quantity
  • Availability
  • Add to Cart